Learn More
Biotang Inc Myostatin Human Recombinant, The ED50 as determined by the inhibition of the prolifiration of MPC-11 cells is < 20ng/ml corresponding to a Specific Activity of 50,000IU/mg.
Shop All BIOTANG Inc Products