Learn More
Biotang Inc Nanog Human Recombinant, produced in E.Coli is a single, non-glycosylated, Polypeptide chain containing 305 amino acids and having a molecular mass of 34.6kDa. The NANOG is fused to an N-terminal TAT (transcriptional activator protein) tag
Catalog No. 50150112 Shop All BIOTANG Inc Products
Encompass_Preferred
Nanog Human Recombinant, produced in E.Coli is a single, non-glycosylated, Polypeptide chain containing 305 amino acids and having a molecular mass of 34.6kDa. The NANOG is fused to an N-terminal TAT (transcriptional activator protein) tag
Catalog No. 50-150-112 Supplier Biotang Inc Supplier No. BTPRO090
Nanog Human Recombinant, produced in E.Coli is a single, non-glycosylated, Polypeptide chain containing 305 amino acids and having a molecular mass of 34.6kDa. The NANOG is fused to an N-terminal TAT (transcriptional activator protein) tag