Learn More
Biotang Inc Noggin Human Recombinant, the ED50 was determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells.
Supplier:  Biotang Inc BTCYT475
Encompass_Preferred
Noggin Human Recombinant, the ED50 was determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells.
Catalog No. 50-150-142