Learn More
Biotang Inc Osteocrin Human Recombinant, The Human Osteocrin His-Tagged Fusion Protein is 13.6 kDa containing 106 amino acid residues of the human Osteocrin and 16 additional amino acid residues -C His Tag
Supplier:  Biotang Inc BTPRO420
Encompass_Preferred
Osteocrin Human Recombinant, The Human Osteocrin His-Tagged Fusion Protein is 13.6 kDa containing 106 amino acid residues of the human Osteocrin and 16 additional amino acid residues -C His Tag
Catalog No. 50-150-170