Learn More
Biotang Inc P-selectin Human Recombinant, is expressed in E. coli containing 566 amino acids 197-761 fused to an amino terminal hexahistidine tag. SELP, GMRP, GRMP, CD62, PSEL, CD62P, GMP140, FLJ45155.
Supplier:  Biotang Inc BTPRO382
Encompass_Preferred
P-selectin Human Recombinant, is expressed in E. coli containing 566 amino acids 197-761 fused to an amino terminal hexahistidine tag. SELP, GMRP, GRMP, CD62, PSEL, CD62P, GMP140, FLJ45155.
Catalog No. 50-150-344