Learn More
Biotang Inc Paxillin Human Recombinant, PXN, Paxillin, FLJ16691. containing 203 amino acids (356-558 a.a.) having a total Mw of 48kDa. Human Paxillin is fused to a GST tag
Supplier:  Biotang Inc BTPRO744
Encompass_Preferred
Paxillin Human Recombinant, PXN, Paxillin, FLJ16691. containing 203 amino acids (356-558 a.a.) having a total Mw of 48kDa. Human Paxillin is fused to a GST tag
Catalog No. 50-150-193