Learn More
Biotang Inc Periostin Human Recombinant, is produced in E. coli, and its molecular weight is 75 kDa protein containing 648 amino acid residues of the human OSF-2 and 23 additional amino acid residues - HisTag, Xa - cleavage site.
Supplier:  Biotang Inc BTCYT452
Encompass_Preferred
Periostin Human Recombinant, is produced in E. coli, and its molecular weight is 75 kDa protein containing 648 amino acid residues of the human OSF-2 and 23 additional amino acid residues - HisTag, Xa - cleavage site.
Catalog No. 50-150-205