Learn More
Biotang Inc Platelet Factor-4 Human Recombinant (CXCL4), PF 4 Human, containing 70 amino acids and having a molecular mass of 7.8 kDa. ED50 is 5-15 ug/ml
Supplier:  Biotang Inc BTCHM350
Encompass_Preferred
Platelet Factor-4 Human Recombinant (CXCL4), PF 4 Human, containing 70 amino acids and having a molecular mass of 7.8 kDa. ED50 is 5-15 ug/ml
Catalog No. 50-150-260