Learn More
Biotang Inc Pleiotrophin Human Recombinant, produced in E. coli, its molecular weight is 17.3 kDa protein containing 136 amino acid residues of the OSF-1 human and 16 additional amino acid residues - HisTag, thrombin cleavage site
Catalog No. 50150263 Shop All BIOTANG Inc Products
Encompass_Preferred
Pleiotrophin Human Recombinant, produced in E. coli, its molecular weight is 17.3 kDa protein containing 136 amino acid residues of the OSF-1 human and 16 additional amino acid residues - HisTag, thrombin cleavage site
Catalog No. 50-150-263 Supplier Biotang Inc Supplier No. BTCYT451
Pleiotrophin Human Recombinant, produced in E. coli, its molecular weight is 17.3 kDa protein containing 136 amino acid residues of the OSF-1 human and 16 additional amino acid residues - HisTag, thrombin cleavage site