Learn More
Biotang Inc Prolactin Human Recombinant, His Tag, containing 199 amino acids fragment (29-227) and having a molecular mass of 23 kDa with an amino-terminal hexahistidine tag.
Supplier:  Biotang Inc BTCYT493
Encompass_Preferred
Prolactin Human Recombinant, His Tag, containing 199 amino acids fragment (29-227) and having a molecular mass of 23 kDa with an amino-terminal hexahistidine tag.
Catalog No. 50-150-293