Learn More
Biotang Inc RAD1 Human Recombinant, HRAD1, REC1, Cell cycle checkpoint protein RAD1,containing 301 amino acids (1-282 a.a.) and having a molecular mass of 33.9 kDa. The RAD1 is fused to 20 amino acid His-Tag at N-terminus
Supplier:  Biotang Inc BTPRO868
Encompass_Preferred
RAD1 Human Recombinant, HRAD1, REC1, Cell cycle checkpoint protein RAD1,containing 301 amino acids (1-282 a.a.) and having a molecular mass of 33.9 kDa. The RAD1 is fused to 20 amino acid His-Tag at N-terminus
Catalog No. 50-150-367