Learn More
Biotang Inc Resistin Human Recombinant, RSTN, XCP1, RETN1.9.9 kDa protein containing 93 amino acid residues.Specific Activity of 100,000IU/mg.
Catalog No. 50150399 Shop All BIOTANG Inc Products
Encompass_Preferred
Resistin Human Recombinant, RSTN, XCP1, RETN1.9.9 kDa protein containing 93 amino acid residues.Specific Activity of 100,000IU/mg.
Catalog No. 50-150-399 Supplier Biotang Inc Supplier No. BTCYT456
Resistin Human Recombinant, RSTN, XCP1, RETN1.9.9 kDa protein containing 93 amino acid residues.Specific Activity of 100,000IU/mg.