Learn More
Biotang Inc Resistin Human Recombinant, His Tag, RSTN, XCP1, RETN1, 9.9 kDa protein containing 93 amino acid residues fused with 4kDa His Tag fusion protein.
Supplier:  Biotang Inc BTCYT256
Encompass_Preferred
Resistin Human Recombinant, His Tag, RSTN, XCP1, RETN1, 9.9 kDa protein containing 93 amino acid residues fused with 4kDa His Tag fusion protein.
Catalog No. 50-150-401