Learn More
Biotang Inc HIV-1 TAT Cys22 Recombinant, HIV-1 TAT Cys22,produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids mutated in the trans activation domain and having chain having a molecular mass of 14.4 kDa.
Catalog No. 50151711 Shop All BIOTANG Inc Products
Encompass_Preferred
HIV-1 TAT Cys22 Recombinant, HIV-1 TAT Cys22,produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids mutated in the trans activation domain and having chain having a molecular mass of 14.4 kDa.
Catalog No. 50-151-711 Supplier Biotang Inc Supplier No. BTHIV136
HIV-1 TAT Cys22 Recombinant, HIV-1 TAT Cys22,produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids mutated in the trans activation domain and having chain having a molecular mass of 14.4 kDa.