Learn More
Biotang Inc Septin-2 Human Recombinant, SEPT2 Human,Septin 2, DIFF6, NEDD5, hNedd5, Pnutl3, NEDD-5. containing 381 amino acids (1-361a.a.) and having a molecular mass of 43.6kDa. SEPT2 is fused to 20 amino acid His Tag at N-terminus
Catalog No. 50150471 Shop All BIOTANG Inc Products
Encompass_Preferred
Septin-2 Human Recombinant, SEPT2 Human,Septin 2, DIFF6, NEDD5, hNedd5, Pnutl3, NEDD-5. containing 381 amino acids (1-361a.a.) and having a molecular mass of 43.6kDa. SEPT2 is fused to 20 amino acid His Tag at N-terminus
Catalog No. 50-150-471 Supplier Biotang Inc Supplier No. BTPRO255
Septin-2 Human Recombinant, SEPT2 Human,Septin 2, DIFF6, NEDD5, hNedd5, Pnutl3, NEDD-5. containing 381 amino acids (1-361a.a.) and having a molecular mass of 43.6kDa. SEPT2 is fused to 20 amino acid His Tag at N-terminus