Learn More
Biotang Inc Sirtuin 2 Human Recombinant, SIRT2 Human, Sirtuin 2, SIR2L2, SIR2-like protein 2, containing 372 amino acids (1-352a.a.) and having a molecular mass of 41.7kDa. SIRT2 is fused to a 20 amino acid His-tag at N-terminus
Supplier:  Biotang Inc BTPRO033
Encompass_Preferred
Sirtuin 2 Human Recombinant, SIRT2 Human, Sirtuin 2, SIR2L2, SIR2-like protein 2, containing 372 amino acids (1-352a.a.) and having a molecular mass of 41.7kDa. SIRT2 is fused to a 20 amino acid His-tag at N-terminus
Catalog No. 50-150-489