Learn More
Biotang Inc Sirtuin 3 Human Recombinant, SIRT3 Human,SIR2-like protein 3, Sir2-like 3, mitochondrial. containing 303 amino acids (118-399a.a.) and having a molecular mass of 33.5kDa. SIRT3 is fused to a 21 amino acid His-tag at N-terminus
Supplier:  Biotang Inc BTPRO462
Encompass_Preferred
Sirtuin 3 Human Recombinant, SIRT3 Human,SIR2-like protein 3, Sir2-like 3, mitochondrial. containing 303 amino acids (118-399a.a.) and having a molecular mass of 33.5kDa. SIRT3 is fused to a 21 amino acid His-tag at N-terminus
Catalog No. 50-150-490