Learn More
Biotang Inc Sirtuin-6 Human Recombinant, SIRT6 Human,containing 375 amino acids (1-355 a.a.) and having a molecular mass of 41 kDa. Recombinant SIRT6 is fused to 20 amino acids His-tag at N-terminus
Supplier:  Biotang Inc BTPRO282
Encompass_Preferred
Sirtuin-6 Human Recombinant, SIRT6 Human,containing 375 amino acids (1-355 a.a.) and having a molecular mass of 41 kDa. Recombinant SIRT6 is fused to 20 amino acids His-tag at N-terminus
Catalog No. 50-150-491