Learn More
Biotang Inc SMAC/DIABLO Human Recombinant, fused to N-terminal T7-Tag produced in E.Coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 22 kDa.
Supplier:  Biotang Inc BTPRO614
Encompass_Preferred
SMAC/DIABLO Human Recombinant, fused to N-terminal T7-Tag produced in E.Coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 22 kDa.
Catalog No. 50-150-492