Learn More
Biotang Inc SOX9 Human Recombinant, containing 195 amino acids (316-510 a.a.) fused to a GST tag and having a total molecular mass of 48kDa.
Supplier:  Biotang Inc BTPRO738
Encompass_Preferred
SOX9 Human Recombinant, containing 195 amino acids (316-510 a.a.) fused to a GST tag and having a total molecular mass of 48kDa.
Catalog No. 50-150-521