Learn More
Biotang Inc SP100 Recombinant Human, Recombinant Sp100 antigen is a full-length cDNA coding having a molecular mass of 54,662 Dalton (pH 5.0). Sp100 protein is fused to a hexa-histidine purification tag.
Supplier:  Biotang Inc BTPRO108
Encompass_Preferred
SP100 Recombinant Human, Recombinant Sp100 antigen is a full-length cDNA coding having a molecular mass of 54,662 Dalton (pH 5.0). Sp100 protein is fused to a hexa-histidine purification tag.
Catalog No. 50-150-523