Learn More
Biotang Inc Ubiquitin Human Recombinant, GST Tag, RPS27A Human, GST. Ubiquitin Human Recombinant GST Tag full length protein is produced in E.Coli, having a molecular weight of 35kDa
Catalog No. 50150708 Shop All BIOTANG Inc Products
Encompass_Preferred
Ubiquitin Human Recombinant, GST Tag, RPS27A Human, GST. Ubiquitin Human Recombinant GST Tag full length protein is produced in E.Coli, having a molecular weight of 35kDa
Catalog No. 50-150-708 Supplier Biotang Inc Supplier No. BTPRO735
Ubiquitin Human Recombinant, GST Tag, RPS27A Human, GST. Ubiquitin Human Recombinant GST Tag full length protein is produced in E.Coli, having a molecular weight of 35kDa