Learn More
Biotang Inc Urokinase Human, Specific Activity 83,700IU/mg. containing 411 amino acids with 12 disulfide bonds. Its molecular weight is 54,000 Dalton.
Supplier:  Biotang Inc BTENZ264
Encompass_Preferred
Urokinase Human, Specific Activity 83,700IU/mg. containing 411 amino acids with 12 disulfide bonds. Its molecular weight is 54,000 Dalton.
Catalog No. 50-150-721