Learn More
Biotang Inc Vaspin Human Recombinant,Vaspin Human Recombinant produced in E.Coli is a single, non-glycosylated, His Tag, polypeptide chain containing 415 amino acids and having a molecular mass of 47 kDa.
Catalog No. 50150752 Shop All BIOTANG Inc Products
Encompass_Preferred
Vaspin Human Recombinant,Vaspin Human Recombinant produced in E.Coli is a single, non-glycosylated, His Tag, polypeptide chain containing 415 amino acids and having a molecular mass of 47 kDa.
Catalog No. 50-150-752 Supplier Biotang Inc Supplier No. BTCYT459
Vaspin Human Recombinant,Vaspin Human Recombinant produced in E.Coli is a single, non-glycosylated, His Tag, polypeptide chain containing 415 amino acids and having a molecular mass of 47 kDa.