Learn More
Biotang Inc Visfatin Human Recombinant, containing 348 amino acids and having 11 N-terminal flag tag The total molecular mass is 39.6 kDa (calculated).
Catalog No. 50150760 Shop All BIOTANG Inc Products
Encompass_Preferred
Visfatin Human Recombinant, containing 348 amino acids and having 11 N-terminal flag tag The total molecular mass is 39.6 kDa (calculated).
Catalog No. 50-150-760 Supplier Biotang Inc Supplier No. BTCYT318
Visfatin Human Recombinant, containing 348 amino acids and having 11 N-terminal flag tag The total molecular mass is 39.6 kDa (calculated).