Learn More
Abcam Recombinant Human ALIX protein, 10UG
Supplier:  Abcam AB13253410UG
Encompass_Preferred
Recombinant Human ALIX protein is a Human Full Length protein, in the 1 to 868 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: AIP1, ALIX, KIAA1375, PDCD6IP, Programmed cell death 6-interacting protein, PDCD6-interacting protein, ALG-2-interacting protein 1, ALG-2-interacting protein X, Hp95
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2007461