Learn More
Biotang Inc L-Asparaginase,Specific Activity 225 IU/mg. produced from E.Coli containing 303 amino acids and having a molecular mass of 31731 Dalton.
Supplier:  Biotang Inc BTENZ287
Encompass_Preferred
L-Asparaginase,Specific Activity 225 IU/mg. produced from E.Coli containing 303 amino acids and having a molecular mass of 31731 Dalton.
Catalog No. 50-150-781