Learn More
Biotang Inc Melanotan-I, MT I, Melanotan-I has the amino acid sequence of Ac-Ser-Tyr-Ser-Nle-Glu-His-D-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2 and a molecular weight of 1645.82 Dalton.
Supplier:  Biotang Inc BTHOR306
Encompass_Preferred
Melanotan-I, MT I, Melanotan-I has the amino acid sequence of Ac-Ser-Tyr-Ser-Nle-Glu-His-D-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2 and a molecular weight of 1645.82 Dalton.
Catalog No. 50-150-791