Learn More
Apexbio Technology LLC REC MU IL-10 HIS-FLAG-1mg
SDP
Catalog No. 5000897640
Encompass_Preferred
Recombinant Mouse IL-10 (His Flag) is a cytokine involved in immunological research originating from the regulatory pathways of mice It plays a crucial role in modulating immune responses by inhibiting pro-inflammatory cytokines and promoting anti-inflammatory signaling This protein functions through the IL-10 receptor complex impacting JAK-STAT pathways to mediate its effects In vitro its activity is typically observed in the nanomolar to low micromolar range Commonly used in various cell models and animal studies this recombinant protein is instrumental in drug screening processes particularly in evaluating anti-inflammatory and immunosuppressive therapies Experimental concentrations typically depend on the specific research design and objectives making it a versatile tool in the investigation of immune system dynamics and therapeutic interventions
Catalog No. 50-008-97640 Supplier Apexbio Technology LLC Supplier No. PM10161
Recombinant Mouse IL-10 (His Flag) is a cytokine involved in immunological research originating from the regulatory pathways of mice It plays a crucial role in modulating immune responses by inhibiting pro-inflammatory cytokines and promoting anti-inflammatory signaling This protein functions through the IL-10 receptor complex impacting JAK-STAT pathways to mediate its effects In vitro its activity is typically observed in the nanomolar to low micromolar range Commonly used in various cell models and animal studies this recombinant protein is instrumental in drug screening processes particularly in evaluating anti-inflammatory and immunosuppressive therapies Experimental concentrations typically depend on the specific research design and objectives making it a versatile tool in the investigation of immune system dynamics and therapeutic interventions