Learn More
Apexbio Technology LLC REC MU IL-13 HIS-FLAG-1mg
SDP
Catalog No. 5000897646
Encompass_Preferred
Recombinant Mouse IL-13 (His Flag) is a cytokine used in biomedical research derived from mouse interleukin-13 and modified with His and Flag tags for purification and detection It plays a crucial role in immunological pathways primarily affecting the differentiation and activity of immune cells IL-13 is known to engage with the IL-13 receptor influencing various signaling cascades that regulate inflammation and immune responses In vitro this recombinant protein displays activity in the nanomolar to low micromolar range depending on the specific assay conditions It is widely applied in cell culture models to study immune modulation and is frequently used in animal models to investigate asthma and allergic responses Additionally it serves in drug screening processes aiming to find novel therapies targeting immune-related disorders Experimental concentrations of Recombinant Mouse IL-13 typically depend on the specific research design and objectives of the study
Catalog No. 50-008-97646 Supplier Apexbio Technology LLC Supplier No. PM10171
Recombinant Mouse IL-13 (His Flag) is a cytokine used in biomedical research derived from mouse interleukin-13 and modified with His and Flag tags for purification and detection It plays a crucial role in immunological pathways primarily affecting the differentiation and activity of immune cells IL-13 is known to engage with the IL-13 receptor influencing various signaling cascades that regulate inflammation and immune responses In vitro this recombinant protein displays activity in the nanomolar to low micromolar range depending on the specific assay conditions It is widely applied in cell culture models to study immune modulation and is frequently used in animal models to investigate asthma and allergic responses Additionally it serves in drug screening processes aiming to find novel therapies targeting immune-related disorders Experimental concentrations of Recombinant Mouse IL-13 typically depend on the specific research design and objectives of the study