Learn More
Apexbio Technology LLC REC MU IL-17 HIS-FLAG-1mg
SDP
Catalog No. 5000897643
Encompass_Preferred
Recombinant Mouse IL-17 (His Flag) is a cytokine used in biomedical research particularly in immunology and inflammation studies Originating from mouse models it plays a crucial role in mediating pro-inflammatory responses by signaling through the IL-17 receptor pathway influencing downstream effects like cytokine production and cell recruitment This protein is commonly tested at nanomolar to low micromolar concentrations for its in vitro activity It is frequently used in cell models to study inflammatory pathways and in animal studies to explore its role in diseases like autoimmune disorders Additionally it serves as an important tool in drug screening for evaluating potential therapeutic interventions targeting IL-17 mediated pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97643 Supplier Apexbio Technology LLC Supplier No. PM10181
Recombinant Mouse IL-17 (His Flag) is a cytokine used in biomedical research particularly in immunology and inflammation studies Originating from mouse models it plays a crucial role in mediating pro-inflammatory responses by signaling through the IL-17 receptor pathway influencing downstream effects like cytokine production and cell recruitment This protein is commonly tested at nanomolar to low micromolar concentrations for its in vitro activity It is frequently used in cell models to study inflammatory pathways and in animal studies to explore its role in diseases like autoimmune disorders Additionally it serves as an important tool in drug screening for evaluating potential therapeutic interventions targeting IL-17 mediated pathways Experimental concentrations typically depend on the specific research design