Learn More
Apexbio Technology LLC REC MU IL-19 HIS-FLAG-1mg
SDP
Catalog No. 5000897645
Encompass_Preferred
Recombinant Mouse IL-19 (His Flag) is a protein used in biomedical research derived from mouse sources It belongs to the IL-10 cytokine family and plays a role in immune response modulation by activating specific signaling pathways such as the JAK-STAT pathway Its activity is commonly noted in the nanomolar to low micromolar range This recombinant protein is frequently utilized in in vitro studies involving cell lines contributing to our understanding of immune signaling and cytokine interactions In vivo it is useful in animal model studies aimed at exploring inflammation-related conditions Additionally it aids in drug screening applications allowing researchers to evaluate potential therapeutic agents targeting inflammatory pathways Experimental concentrations typically depend on the specific research design ensuring precision and relevance to the study s objectives
Catalog No. 50-008-97645 Supplier Apexbio Technology LLC Supplier No. PM10231
Recombinant Mouse IL-19 (His Flag) is a protein used in biomedical research derived from mouse sources It belongs to the IL-10 cytokine family and plays a role in immune response modulation by activating specific signaling pathways such as the JAK-STAT pathway Its activity is commonly noted in the nanomolar to low micromolar range This recombinant protein is frequently utilized in in vitro studies involving cell lines contributing to our understanding of immune signaling and cytokine interactions In vivo it is useful in animal model studies aimed at exploring inflammation-related conditions Additionally it aids in drug screening applications allowing researchers to evaluate potential therapeutic agents targeting inflammatory pathways Experimental concentrations typically depend on the specific research design ensuring precision and relevance to the study s objectives