Learn More
Apexbio Technology LLC REC MU IL-2 HIS-STREP-1mg
SDP
Catalog No. 5000897661
Encompass_Preferred
Recombinant Mouse IL-2 (His Strep) is a critical component in the field of immunology research often used to investigate cytokine signaling pathways Originating from mouse sources this cytokine is engineered to include histidine and streptavidin tags for facilitating purification and detection It primarily targets the interleukin-2 receptor playing a pivotal role in the proliferation and differentiation of T-cells and natural killer cells This protein exhibits in vitro activity typically in the nanomolar to micromolar range Recombinant Mouse IL-2 is commonly applied in various cell models animal studies and drug screening processes to explore immune response mechanisms Experimental concentrations typically depend on the specific research design allowing for adaptability across diverse experimental setups
Catalog No. 50-008-97661 Supplier Apexbio Technology LLC Supplier No. PM10131
Recombinant Mouse IL-2 (His Strep) is a critical component in the field of immunology research often used to investigate cytokine signaling pathways Originating from mouse sources this cytokine is engineered to include histidine and streptavidin tags for facilitating purification and detection It primarily targets the interleukin-2 receptor playing a pivotal role in the proliferation and differentiation of T-cells and natural killer cells This protein exhibits in vitro activity typically in the nanomolar to micromolar range Recombinant Mouse IL-2 is commonly applied in various cell models animal studies and drug screening processes to explore immune response mechanisms Experimental concentrations typically depend on the specific research design allowing for adaptability across diverse experimental setups