Learn More
Apexbio Technology LLC REC MU IL-4 HIS-STREP-1mg
SDP
Catalog No. 5000897657
Encompass_Preferred
Recombinant Mouse IL-4 (His Strep) is a cytokine used in biomedical research originating from mouse models to study immune responses It functions by binding to the IL-4 receptor activating the JAK-STAT pathway and influencing the differentiation of naive helper T cells into Th2 cells Its activity is typically within the nanomolar to low micromolar range depending on the assay Common applications include studies in cell models to investigate immune cell behavior animal studies to understand systemic immune responses and drug screening to evaluate potential immunomodulatory agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97657 Supplier Apexbio Technology LLC Supplier No. PM10141
Recombinant Mouse IL-4 (His Strep) is a cytokine used in biomedical research originating from mouse models to study immune responses It functions by binding to the IL-4 receptor activating the JAK-STAT pathway and influencing the differentiation of naive helper T cells into Th2 cells Its activity is typically within the nanomolar to low micromolar range depending on the assay Common applications include studies in cell models to investigate immune cell behavior animal studies to understand systemic immune responses and drug screening to evaluate potential immunomodulatory agents Experimental concentrations typically depend on the specific research design