Learn More
Apexbio Technology LLC REC MU M-CSF HIS-FLAG-1mg
SDP
Catalog No. 5000897662
Encompass_Preferred
Recombinant Mouse M-CSF/CSF1 (His Flag) is a critical cytokine involved in the regulation of macrophage lineage cells originating from murine sources It functions by binding to its receptor CSF1R on the cell surface triggering downstream signaling pathways crucial for survival proliferation and differentiation of macrophages In vitro activities are often observed in the nanomolar to low micromolar range making it a valuable tool in research It is widely used in cell models animal studies and drug screening to explore macrophage-related functions and therapeutic interventions Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97662 Supplier Apexbio Technology LLC Supplier No. PM10191
Recombinant Mouse M-CSF/CSF1 (His Flag) is a critical cytokine involved in the regulation of macrophage lineage cells originating from murine sources It functions by binding to its receptor CSF1R on the cell surface triggering downstream signaling pathways crucial for survival proliferation and differentiation of macrophages In vitro activities are often observed in the nanomolar to low micromolar range making it a valuable tool in research It is widely used in cell models animal studies and drug screening to explore macrophage-related functions and therapeutic interventions Experimental concentrations typically depend on the specific research design