Learn More
Biotang Inc Epidermal Growth Factor Mouse Recombinant, EGF Mouse, containing 53 amino acids including 3 intramolecular disulfide-bonds and having a molecular mass of 6 kDa. Source Escherichia Coli.
Catalog No. 50150824 Shop All BIOTANG Inc Products
Encompass_Preferred
Epidermal Growth Factor Mouse Recombinant, EGF Mouse, containing 53 amino acids including 3 intramolecular disulfide-bonds and having a molecular mass of 6 kDa. Source Escherichia Coli.
Catalog No. 50-150-824 Supplier Biotang Inc Supplier No. BTCYT326
Epidermal Growth Factor Mouse Recombinant, EGF Mouse, containing 53 amino acids including 3 intramolecular disulfide-bonds and having a molecular mass of 6 kDa. Source Escherichia Coli.