Learn More
Biotang Inc Pegylated Mouse Leptin Recombinant, Leptin Mouse, PEG,containing 146 aa and an additional Ala at N-terminus.contains PEG 20 kDa at its N-terminus and having a molecular mass of 35.6 kDa as determined by mass spectrometry.
Supplier:  Biotang Inc BTCYT591
Encompass_Preferred
Pegylated Mouse Leptin Recombinant, Leptin Mouse, PEG,containing 146 aa and an additional Ala at N-terminus.contains PEG 20 kDa at its N-terminus and having a molecular mass of 35.6 kDa as determined by mass spectrometry.
Catalog No. 50-150-950