Learn More
Biotang Inc RELM-Gamma Mouse Recombinant, RELM g Mouse, having a molecular weight of 11 kDa containing 86 amino acid residues of the RELM-gamma Mouse and 16 additional amino acid residues ~C HisTag
Supplier:  Biotang Inc BTCYT455
Encompass_Preferred
RELM-Gamma Mouse Recombinant, RELM g Mouse, having a molecular weight of 11 kDa containing 86 amino acid residues of the RELM-gamma Mouse and 16 additional amino acid residues ~C HisTag
Catalog No. 50-150-910