Learn More
Biotang Inc Resistin Mouse Recombinant, with signal sequence of phage fd (21aa) and C-terminal fusion of flagTag (10aa). Resistin Mouse Recombinant Flag-Tagged Fusion Protein is 13.7 kDa protein containing 93 amino acid residues of the Resistin Mouse
Supplier:  Biotang Inc BTCYT457
Encompass_Preferred
Resistin Mouse Recombinant, with signal sequence of phage fd (21aa) and C-terminal fusion of flagTag (10aa). Resistin Mouse Recombinant Flag-Tagged Fusion Protein is 13.7 kDa protein containing 93 amino acid residues of the Resistin Mouse
Catalog No. 50-150-911