Learn More
Biotang Inc Visfatin Mouse Recombinant, containing His tag and having 511 amino acids. The total molecular mass is 57 kDa. Source Escherichia Coli.
Catalog No. 50150929 Shop All BIOTANG Inc Products
Encompass_Preferred
Visfatin Mouse Recombinant, containing His tag and having 511 amino acids. The total molecular mass is 57 kDa. Source Escherichia Coli.
Catalog No. 50-150-929 Supplier Biotang Inc Supplier No. BTCYT447
Visfatin Mouse Recombinant, containing His tag and having 511 amino acids. The total molecular mass is 57 kDa. Source Escherichia Coli.