Learn More
Bio X Cell IgG4 (S228P) Isotype Control, Anti-HEL, 100 MG
Supplier: Bio X Cell CP147R100MG
This human IgG4 S228P isotype control antibody reacts with hen egg lysozyme and has low or no specific binding to any human sample. The S228P mutation is included to prevent IgG4 Fab exchange. This is a recombinant human IgG4 antibody produced in CHO cells.
- Isotype: Human IgG4, κ
- Mutations: S228P
- Immunogen: Hen egg lysozyme (HEL)
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein A
- Aggregation: <5%
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.