Learn More
Biotang Inc Leptin Ovine Recombinant, MTS tag, containing 157 amino acids and having a molecular mass of 17.5 kDa. The membrane translocating sequence Tag is composed of 10 amino acids at the N-terminus.
Catalog No. 50150944 Shop All BIOTANG Inc Products
Encompass_Preferred
Leptin Ovine Recombinant, MTS tag, containing 157 amino acids and having a molecular mass of 17.5 kDa. The membrane translocating sequence Tag is composed of 10 amino acids at the N-terminus.
Catalog No. 50-150-944 Supplier Biotang Inc Supplier No. BTCYT531
Leptin Ovine Recombinant, MTS tag, containing 157 amino acids and having a molecular mass of 17.5 kDa. The membrane translocating sequence Tag is composed of 10 amino acids at the N-terminus.