Learn More
Biotang Inc Pfu-DNA Polymerase Recombinant, Source Escherichia Coli. Pfu DNA Polymerase is a thermo-stable enzyme having a Mw of about 90kDa.
Catalog No. 50150954 Shop All BIOTANG Inc Products
Encompass_Preferred
Pfu-DNA Polymerase Recombinant, Source Escherichia Coli. Pfu DNA Polymerase is a thermo-stable enzyme having a Mw of about 90kDa.
Catalog No. 50-150-954 Supplier Biotang Inc Supplier No. BTENZ265
Pfu-DNA Polymerase Recombinant, Source Escherichia Coli. Pfu DNA Polymerase is a thermo-stable enzyme having a Mw of about 90kDa.