Learn More
Biotang Inc Protein A/G Recombinant, Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins.
Supplier:  Biotang Inc BTPRO646
Encompass_Preferred
Protein A/G Recombinant, Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins.
Catalog No. 50-150-983