Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rec Protein A

Catalog No. 101100
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-110-0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-110-0 Supplier Invitrogen™ Supplier No. 101100
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rec Protein A

This recombinant protein A from Staphylococcus aureus Cowan I was expressed in Bacillus. The gene has been altered to truncate the protein A at the carboxyl terminal. This does not affect the binding characteristics.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C
Shipping Condition Wet Ice
Label or Dye Unconjugated
Type Protein A
Quantity 5 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.