Learn More
Biotang Inc Epidermal Growth Factor Rat Recombinant, EGF Rat, produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 53 amino acids and having a molecular mass of 6151 Dalton.
Supplier:  Biotang Inc BTCYT669
Encompass_Preferred
Epidermal Growth Factor Rat Recombinant, EGF Rat, produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 53 amino acids and having a molecular mass of 6151 Dalton.
Catalog No. 50-151-005