Learn More
Biotang Inc Resistin Rat Recombinant, Resistin Rat Recombinant His-Tagged Fusion Protein is an 11.9 kDa protein containing 94 amino acid residues of the Resistin Rat and 16 additional amino acid residues
Catalog No. 50151042 Shop All BIOTANG Inc Products
Encompass_Preferred
Resistin Rat Recombinant, Resistin Rat Recombinant His-Tagged Fusion Protein is an 11.9 kDa protein containing 94 amino acid residues of the Resistin Rat and 16 additional amino acid residues
Catalog No. 50-151-042 Supplier Biotang Inc Supplier No. BTCYT458
Resistin Rat Recombinant, Resistin Rat Recombinant His-Tagged Fusion Protein is an 11.9 kDa protein containing 94 amino acid residues of the Resistin Rat and 16 additional amino acid residues