Learn More
Biotang Inc Secretagogin Rat Recombinant, SCGN Rat, produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 286 amino acids and having a molecular mass of 33.3 kDa. The Rat SCGN is fused to a 10 a.a. His tag at N-Terminus.
Catalog No. 50151044 Shop All BIOTANG Inc Products
Encompass_Preferred
Secretagogin Rat Recombinant, SCGN Rat, produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 286 amino acids and having a molecular mass of 33.3 kDa. The Rat SCGN is fused to a 10 a.a. His tag at N-Terminus.
Catalog No. 50-151-044 Supplier Biotang Inc Supplier No. BTPRO657
Secretagogin Rat Recombinant, SCGN Rat, produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 286 amino acids and having a molecular mass of 33.3 kDa. The Rat SCGN is fused to a 10 a.a. His tag at N-Terminus.