Learn More
Abcam Recombinant Rubella Virus capsid protein, 200UG
Supplier:  Abcam AB43034200UG
Encompass_Preferred
Recombinant Rubella Virus capsid protein is a Rubella virus strain Therien Fragment protein, expressed in Escherichia coli, with >95% purity and suitable for ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Structural polyprotein, p110
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2061253