Learn More
Biotang Inc GST-Alpha Spectrin Recombinant, containing repeat 17 and the ubiquitination target lysine 27. The total molecular weight of the fusion protein is 52.4 kDa.
Supplier:  Biotang Inc BTPRO404
Encompass_Preferred
GST-Alpha Spectrin Recombinant, containing repeat 17 and the ubiquitination target lysine 27. The total molecular weight of the fusion protein is 52.4 kDa.
Catalog No. 50-151-066